missing translation for 'onlineSavingsMsg'
Learn More

6-Phenyl-2-thiouracil 98.0+%, TCI America™

Catalog No. P02365G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P02365G 5 g
P02361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P02365G Supplier TCI America Supplier No. P02365G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 36822-11-4
Molecular Formula C10H8N2OS
Molecular Weight (g/mol) 204.247
MDL Number MFCD00059166
InChI Key XEKNACRTWJHOCE-UHFFFAOYSA-N
PubChem CID 697057
IUPAC Name 6-phenyl-2-sulfanylidene-1H-pyrimidin-4-one
SMILES C1=CC=C(C=C1)C2=CC(=O)NC(=S)N2

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 204.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Phenyl-2-thiouracil