missing translation for 'onlineSavingsMsg'
Learn More

6-Phenyl-2-hexyne, 99%

Catalog No. AAB2001903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2001903 1 g
AAB2001906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2001903 Supplier Thermo Scientific Chemicals Supplier No. B2001903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 34298-75-4 | C12H14 | 158.244 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34298-75-4
Molecular Formula C12H14
Molecular Weight (g/mol) 158.244
MDL Number MFCD00026982
InChI Key BYQGPSNMZZGRQP-UHFFFAOYSA-N
Synonym 6-phenyl-2-hexyne, hex-4-yn-1-ylbenzene, 1-phenyl-4-hexyne, acmc-20alxr, methyl 3-phenylpropyl acetylene
PubChem CID 3251198
IUPAC Name hex-4-ynylbenzene
SMILES CC#CCCCC1=CC=CC=C1

Specifications

Quantity 1 g
Formula Weight 158.25
Percent Purity 99%
Chemical Name or Material 6-Phenyl-2-hexyne
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only