missing translation for 'onlineSavingsMsg'
Learn More

6-Phenyl-1-hexanol, 97%

Catalog No. AC308470010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC308470010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC308470010 Supplier Thermo Scientific Chemicals Supplier No. 308470010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2430-16-2 | C12H18O | 178.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2430-16-2
Molecular Formula C12H18O
Molecular Weight (g/mol) 178.28
MDL Number MFCD00014063
InChI Key FDXBUMXUJRZANT-UHFFFAOYSA-N
Synonym 6-phenyl-1-hexanol, benzenehexanol, 6-phenylhexanol, 6-phenyl-n-hexanol, zlchem 717, acmc-20alxt, 6-phenyl hexanol-1, 6-phenyl-hexan-1-ol
PubChem CID 520110
IUPAC Name 6-phenylhexan-1-ol
SMILES OCCCCCCC1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.9500g/mL
Boiling Point 154°C to 155°C (11.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H5(CH2)6OH
Refractive Index 1.5100 to 1.512
Quantity 1 g
Specific Gravity 0.95
Formula Weight 178.28
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 6-Phenyl-1-hexanol, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only