missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Phenoxynicotinaldehyde, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC19604DA
| Quantity | 1g |
|---|---|
| Packaging | Amber Glass Bottle |
Chemical Identifiers
| 173282-69-4 | |
| 199.209 | |
| SOPOQXNWJNVZMI-UHFFFAOYSA-N | |
| 2776500 | |
| C1=CC=C(C=C1)OC2=NC=C(C=C2)C=O |
| C12H9NO2 | |
| MFCD02681950 | |
| 6-phenoxynicotinaldehyde, 6-phenoxypyridine-3-carboxaldehyde, 2-phenoxypyridine-5-carboxaldehyde, 3-pyridinecarboxaldehyde,6-phenoxy, 5-formyl-2-phenoxypyridine, 6-phenoxy-3-pyridinecarboxaldehyde, 6-phenoxy-pyridine-3-carbaldehyde, 3-pyridinecarboxaldehyde, 6-phenoxy | |
| 6-phenoxypyridine-3-carbaldehyde |
Specifications
| 6-Phenoxynicotinaldehyde | |
| 96.5 | |
| 97% | |
| C12H9NO2 | |
| MFCD02681950 | |
| SOPOQXNWJNVZMI-UHFFFAOYSA-N | |
| 6-phenoxypyridine-3-carbaldehyde | |
| 2776500 | |
| 97% |
| 173282-69-4 | |
| 100.0 | |
| Amber Glass Bottle | |
| 1g | |
| 6-phenoxynicotinaldehyde, 6-phenoxypyridine-3-carboxaldehyde, 2-phenoxypyridine-5-carboxaldehyde, 3-pyridinecarboxaldehyde,6-phenoxy, 5-formyl-2-phenoxypyridine, 6-phenoxy-3-pyridinecarboxaldehyde, 6-phenoxy-pyridine-3-carbaldehyde, 3-pyridinecarboxaldehyde, 6-phenoxy | |
| C1=CC=C(C=C1)OC2=NC=C(C=C2)C=O | |
| 199.209 | |
| 199.21 |