missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Oxo-1,6-dihydropyridazine-3-carboxylic acid monohydrate, 97%, Thermo Scientific™
$61.32 - $61.32
Chemical Identifiers
| CAS | 37972-69-3 |
|---|---|
| Molecular Formula | C5H3N2O3 |
| Molecular Weight (g/mol) | 139.09 |
| MDL Number | MFCD09064936 |
| InChI Key | GIFSROMQVPUQFK-UHFFFAOYSA-M |
| Synonym | 6-hydroxypyridazine-3-carboxylic acid, 6-hydroxy-3-pyridazinecarboxylic acid, 6-oxo-1,6-dihydropyridazine-3-carboxylic acid, 6-oxo-1,6-dihydro-pyridazine-3-carboxylic acid, 3-hydroxypyridazine-6-carboxylic acid, 3-pyridazinecarboxylic acid, 1,6-dihydro-6-oxo, 6-hydroxy-pyridazine-3-carboxylic acid, 6-oxo-1,6-dihydro-3-pyridazinecarboxylic acid, 3-pyridazinecarboxylicacid, 1,6-dihydro-6-oxo, 6-oxohydropyridazine-3-carboxylic acid |
| PubChem CID | 305970 |
| IUPAC Name | 6-oxo-1H-pyridazine-3-carboxylic acid |
| SMILES | [O-]C(=O)C1=NNC(=O)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
BTB07063DA
|
Thermo Scientific Chemicals
BTB07063DA |
1 g | Amber Glass Bottle |
Each for $61.32
|
|
||||
Chemical Identifiers
| 37972-69-3 | |
| 139.09 | |
| GIFSROMQVPUQFK-UHFFFAOYSA-M | |
| 305970 | |
| [O-]C(=O)C1=NNC(=O)C=C1 |
| C5H3N2O3 | |
| MFCD09064936 | |
| 6-hydroxypyridazine-3-carboxylic acid, 6-hydroxy-3-pyridazinecarboxylic acid, 6-oxo-1,6-dihydropyridazine-3-carboxylic acid, 6-oxo-1,6-dihydro-pyridazine-3-carboxylic acid, 3-hydroxypyridazine-6-carboxylic acid, 3-pyridazinecarboxylic acid, 1,6-dihydro-6-oxo, 6-hydroxy-pyridazine-3-carboxylic acid, 6-oxo-1,6-dihydro-3-pyridazinecarboxylic acid, 3-pyridazinecarboxylicacid, 1,6-dihydro-6-oxo, 6-oxohydropyridazine-3-carboxylic acid | |
| 6-oxo-1H-pyridazine-3-carboxylic acid |
Specifications
| 37972-69-3 | |
| Amber Glass Bottle | |
| MFCD09064936 | |
| 6-hydroxypyridazine-3-carboxylic acid, 6-hydroxy-3-pyridazinecarboxylic acid, 6-oxo-1,6-dihydropyridazine-3-carboxylic acid, 6-oxo-1,6-dihydro-pyridazine-3-carboxylic acid, 3-hydroxypyridazine-6-carboxylic acid, 3-pyridazinecarboxylic acid, 1,6-dihydro-6-oxo, 6-hydroxy-pyridazine-3-carboxylic acid, 6-oxo-1,6-dihydro-3-pyridazinecarboxylic acid, 3-pyridazinecarboxylicacid, 1,6-dihydro-6-oxo, 6-oxohydropyridazine-3-carboxylic acid | |
| [O-]C(=O)C1=NNC(=O)C=C1 | |
| 139.09 | |
| 158.12 | |
| 6-Hydroxy-3-pyridazinecarboxylic acid monohydrate |
| 97% | |
| C5H3N2O3 | |
| 1 g | |
| GIFSROMQVPUQFK-UHFFFAOYSA-M | |
| 6-oxo-1H-pyridazine-3-carboxylic acid | |
| 305970 | |
| 97% |
Safety and Handling
Warning