missing translation for 'onlineSavingsMsg'
Learn More

6-Oxo-1,6-dihydropyridazine-3-carboxylic acid monohydrate, 97%, Thermo Scientific™

Catalog No. BTB07063DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB07063DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTB07063DA Supplier Thermo Scientific Chemicals Supplier No. BTB07063DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 37972-69-3
Molecular Formula C5H3N2O3
Molecular Weight (g/mol) 139.09
MDL Number MFCD09064936
InChI Key GIFSROMQVPUQFK-UHFFFAOYSA-M
Synonym 6-hydroxypyridazine-3-carboxylic acid, 6-hydroxy-3-pyridazinecarboxylic acid, 6-oxo-1,6-dihydropyridazine-3-carboxylic acid, 6-oxo-1,6-dihydro-pyridazine-3-carboxylic acid, 3-hydroxypyridazine-6-carboxylic acid, 3-pyridazinecarboxylic acid, 1,6-dihydro-6-oxo, 6-hydroxy-pyridazine-3-carboxylic acid, 6-oxo-1,6-dihydro-3-pyridazinecarboxylic acid, 3-pyridazinecarboxylicacid, 1,6-dihydro-6-oxo, 6-oxohydropyridazine-3-carboxylic acid
PubChem CID 305970
IUPAC Name 6-oxo-1H-pyridazine-3-carboxylic acid
SMILES [O-]C(=O)C1=NNC(=O)C=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 158.12
Percent Purity 97%
Chemical Name or Material 6-Hydroxy-3-pyridazinecarboxylic acid monohydrate
Hazard Category Warning