missing translation for 'onlineSavingsMsg'
Learn More

6-Oxa-1-azaspiro[3.3]heptane hemioxalate, 95%

Catalog No. p-7038647
Change view
Click to view available options
Quantity:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH57658MD 250 mg
AAH57658ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH57658MD Supplier Thermo Scientific Chemicals Supplier No. H57658MD

CAS: 1380571-72-1 | C12H20N2O6 | 288.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Incompatible with strong oxidizing agents, strong acids and bases.

Chemical Identifiers

CAS 1380571-72-1
Molecular Formula C12H20N2O6
Molecular Weight (g/mol) 288.3
MDL Number MFCD22373634
InChI Key UMKDEMSXCWMHRS-UHFFFAOYSA-N
Synonym 6-oxa-1-azaspiro 3.3 heptane oxalate 2:1, 6-oxa-1-azaspiro 3.3 heptane hemioxalate, 6-oxa-1-azaspiro 3.3 heptane oxalate 2, bis 6-oxa-1-azaspiro 3.3 heptane oxalate, 6-oxa-1-azaspiro 3.3 heptane,ethanedioate 2:1, oxalic acid-6-oxa-1-azaspiro 3.3 heptane 1/2, bis 6-oxa-1-azaspiro 3.3 heptane ; oxalic acid
PubChem CID 71299818
IUPAC Name 6-oxa-1-azaspiro[3.3]heptane;oxalic acid
SMILES C1CNC12COC2.C1CNC12COC2.C(=O)(C(=O)O)O

Specifications

Quantity 250 mg
Formula Weight 144.15
Percent Purity 95%
Chemical Name or Material 6-Oxa-1-azaspiro[3.3]heptane hemioxalate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only