missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Nitroindoline 97.0+%, TCI America™
$37.29 - $137.94
Chemical Identifiers
| CAS | 19727-83-4 |
|---|---|
| Molecular Formula | C8H8N2O2 |
| Molecular Weight (g/mol) | 164.16 |
| MDL Number | MFCD00005710 |
| InChI Key | LTNYDSMDSLOMSM-UHFFFAOYSA-N |
| Synonym | 6-nitroindoline, indoline, 6-nitro, 1h-indole, 2,3-dihydro-6-nitro, 6-nitro-indoline, 2,3-dihydro-6-nitro-1h-indole, ccris 3302, zlchem 357, pubchem7443 |
| PubChem CID | 29757 |
| IUPAC Name | 6-nitro-2,3-dihydro-1H-indole |
| SMILES | [O-][N+](=O)C1=CC=C2CCNC2=C1 |
Chemical Identifiers
| 19727-83-4 | |
| 164.16 | |
| LTNYDSMDSLOMSM-UHFFFAOYSA-N | |
| 29757 | |
| [O-][N+](=O)C1=CC=C2CCNC2=C1 |
| C8H8N2O2 | |
| MFCD00005710 | |
| 6-nitroindoline, indoline, 6-nitro, 1h-indole, 2,3-dihydro-6-nitro, 6-nitro-indoline, 2,3-dihydro-6-nitro-1h-indole, ccris 3302, zlchem 357, pubchem7443 | |
| 6-nitro-2,3-dihydro-1H-indole |
Specifications
| 19727-83-4 | |
| C8H8N2O2 | |
| 1 g | |
| LTNYDSMDSLOMSM-UHFFFAOYSA-N | |
| 6-nitro-2,3-dihydro-1H-indole | |
| 29757 | |
| ≥97.0% (GC,T) | |
| 6-Nitroindoline |
| 67°C | |
| MFCD00005710 | |
| 6-nitroindoline, indoline, 6-nitro, 1h-indole, 2,3-dihydro-6-nitro, 6-nitro-indoline, 2,3-dihydro-6-nitro-1h-indole, ccris 3302, zlchem 357, pubchem7443 | |
| [O-][N+](=O)C1=CC=C2CCNC2=C1 | |
| 164.16 | |
| 164.16 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : NM1950000
TSCA : No