missing translation for 'onlineSavingsMsg'
Learn More

6-Nitrocoumarin 98.0+%, TCI America™

Catalog No. p-7134028
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
N09235G 5 g
N092325G 25 g
2 options

Catalog No. N09235G

Supplier: TCI America N09235G

Only null left

Chemical Identifiers

CAS 2725-81-7
Molecular Formula C9H5NO4
Molecular Weight (g/mol) 191.142
MDL Number MFCD00016973
InChI Key RMERXEXZXIVNBF-UHFFFAOYSA-N
Synonym 6-nitrocoumarin, 6-nitro-2h-chromen-2-one, 6-nitro-2-benzopyrone, 2h-1-benzopyran-2-one, 6-nitro, acmc-20abd6, maybridge1_002226, amtgc084, 6-nitro-2h-chromen-2-one #, 2h-1-benzopyran-2-one,6-nitro, 6-hydroxy oxido amino-2h-chromen-2-one
PubChem CID 75944
IUPAC Name 6-nitrochromen-2-one
SMILES C1=CC2=C(C=CC(=O)O2)C=C1[N+](=O)[O-]

Specifications

Melting Point 191°C
Color White-Yellow
Quantity 5 g
Formula Weight 191.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Nitrocoumarin