missing translation for 'onlineSavingsMsg'
Learn More

6-Nitrocoumarin, 98+%

Catalog No. p-7022159
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1479206 5 g
AAA1479214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1479206 Supplier Thermo Scientific Chemicals Supplier No. A1479206

CAS: 2725-81-7 | C9H5NO4 | 191.142 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2725-81-7
Molecular Formula C9H5NO4
Molecular Weight (g/mol) 191.142
MDL Number MFCD00016973
InChI Key RMERXEXZXIVNBF-UHFFFAOYSA-N
PubChem CID 75944
IUPAC Name 6-nitrochromen-2-one
SMILES C1=CC2=C(C=CC(=O)O2)C=C1[N+](=O)[O-]

Specifications

Melting Point 186°C to 190°C
Odor Odorless
Quantity 5 g
Beilstein 166079
Synonym 6-nitrocoumarin, 6-nitro-2h-chromen-2-one, 6-nitro-2-benzopyrone, 2h-1-benzopyran-2-one, 6-nitro, acmc-20abd6, maybridge1_002226, amtgc084, 6-nitro-2h-chromen-2-one #, 2h-1-benzopyran-2-one,6-nitro, 6-hydroxy oxido amino-2h-chromen-2-one
Formula Weight 191.14
Percent Purity ≥98%
Chemical Name or Material 6-Nitrocoumarin
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 220-341-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only