missing translation for 'onlineSavingsMsg'
Learn More

6-Nitro-2(3H)-benzoxazolone, 98%

Catalog No. AAH2779603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2779603 1 g
AAH2779606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2779603 Supplier Thermo Scientific Chemicals Supplier No. H2779603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4694-91-1 | C7H4N2O4 | 180.119 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4694-91-1
Molecular Formula C7H4N2O4
Molecular Weight (g/mol) 180.119
MDL Number MFCD00463755
InChI Key JGYJZHYTADCWIK-UHFFFAOYSA-N
Synonym 6-nitrobenzo d oxazol-2 3h-one, 6-nitro-2-benzoxazolinone, 2 3h-benzoxazolone, 6-nitro, 6-nitrobenzoxazolinone, 6-nitro benzoxazolinone, 6-nitro-3h-benzooxazol-2-one, 6-nitrobenzoxazol-2 3h-one, 6-nitrobenzoxazolone, 6-nitrobenzoxazole-2 3h-one, 6-nitro-1,3-benzoxazol-2 3h-one
PubChem CID 78419
IUPAC Name 6-nitro-3H-1,3-benzoxazol-2-one
SMILES C1=CC2=C(C=C1[N+](=O)[O-])OC(=O)N2

Specifications

Melting Point 244°C to 249°C
Quantity 1 g
Formula Weight 180.12
Percent Purity 98%
Chemical Name or Material 6-Nitro-2(3H)-benzoxazolone
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 225-157-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only