missing translation for 'onlineSavingsMsg'
Learn More

6-Morpholino-3-pyridinyl isothiocyanate, 97%, Thermo Scientific™

Numéro de catalogue. CC17507DA
Modifier l'affichage
Click to view available options
Quantité:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC17507DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC17507DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC17507DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 52024-29-0
Formule moléculaire C10H11N3OS
Poids moléculaire (g/mol) 221.278
Numéro MDL MFCD03086114
Clé InChI VLZWMPRZGPULIU-UHFFFAOYSA-N
Synonyme 4-5-isothiocyanatopyridin-2-yl morpholine, 6-morpholino-3-pyridinyl isothiocyanate, 6-morpholino-3-pyridinylisothiocyanate, 4-5-isothiocyanato-2-pyridyl morpholine, 4-5-isothiocyatopyridin-2-yl morpholine, 6-morpholin-4-ylpyridin-3-isothiocyanate, 4-5-isothiocyanato-2-pyridinyl morpholine, 5-isothiocyanato-2-morpholin-4-yl pyridine, morpholine,4-5-isothiocyanato-2-pyridinyl, 6-morpholin-4-yl pyridin-3-yl isothiocyanate
PubChem CID 2776464
Nom de l’IUPAC 4-(5-isothiocyanatopyridine-2-yl)morpholine
SOURIRES C1COCCN1C2=NC=C(C=C2)N=C=S

Spécifications

Nom ou substance chimique 6-Morpholino-3-pyridinyl isothiocyanate
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Bouteille en verre ambrée
Quantité 1 g
Poids de formule 221.28
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.