missing translation for 'onlineSavingsMsg'
Learn More

6-morpholin-4-ylpyridazine-3-carboxylic acid hydrochloride hydrate, Thermo Scientific™

Catalog No. p-4391436
Change view
Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC77751CB 250mg Amber Glass Bottle
CC77751DA 1g Amber Glass Bottle
CC77751DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC77751CB Supplier Thermo Scientific Chemicals Supplier No. CC77751CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1192758-40-9
Molecular Formula C9H14ClN3O4
Molecular Weight (g/mol) 263.678
InChI Key VARLCSQEQQKXKH-UHFFFAOYSA-N
Synonym 6-morpholinopyridazine-3-carboxylic acid hydrochloride hydrate, 6-morpholin-4-ylpyridazine-3-carboxylic acid hydrochloride hydrate, 6-morpholin-4-yl pyridazine-3-carboxylic acid hydrate hydrochloride
PubChem CID 53444630
IUPAC Name 6-morpholin-4-ylpyridazine-3-carboxylic acid;hydrate;hydrochloride
SMILES C1COCCN1C2=NN=C(C=C2)C(=O)O.O.Cl

Specifications

Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 245.67
Percent Purity 97%
Chemical Name or Material 6-morpholin-4-ylpyridazine-3-carboxylic acid hydrochloride hydrate