missing translation for 'onlineSavingsMsg'
Learn More

6-Methyluracil 99.0+%, TCI America™

Catalog No. p-7132182
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M045425G 25 g
M0454500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M045425G Supplier TCI America Supplier No. M045425G
Only null left

Chemical Identifiers

CAS 626-48-2
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.12
MDL Number MFCD00006028
InChI Key SHVCSCWHWMSGTE-UHFFFAOYSA-N
Synonym 6-methyluracil, 2,4-dihydroxy-6-methylpyrimidine, pseudothymine, 6-methylpyrimidine-2,4 1h,3h-dione, 4-methyluracil, 6-methylpyrimidine-2,4-diol, 2,4 1h,3h-pyrimidinedione, 6-methyl, uracil, 6-methyl, 6-methyl-2,4 1h,3h-pyrimidinedione, 2,4-pyrimidinediol, 6-methyl
PubChem CID 12283
ChEBI CHEBI:74733
IUPAC Name 6-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES CC1=CC(=O)NC(=O)N1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 126.12
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methyluracil
EINECSNumber (5)-0934
RTECSNumber YR0700000
TSCA Yes