missing translation for 'onlineSavingsMsg'
Learn More

6-Methylquinoline 98.0+%, TCI America™

Catalog No. p-7132156
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M04165G 5 g
M041625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M04165G Supplier TCI America Supplier No. M04165G
Only null left

Chemical Identifiers

CAS 91-62-3
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.19
MDL Number MFCD00006804
InChI Key LUYISICIYVKBTA-UHFFFAOYSA-N
Synonym quinoline, 6-methyl, p-toluquinoline, 6-methyl-quinoline, tolliquinoline, p, ccris 407, fema no. 2744, quinoline, 6-methyl-8ci,9ci, dsstox_cid_887, dsstox_rid_75844, dsstox_gsid_20887
PubChem CID 7059
IUPAC Name 6-methylquinoline
SMILES CC1=CC=C2N=CC=CC2=C1

Specifications

Color Yellow
Boiling Point 259°C
Quantity 5 g
Formula Weight 143.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 6-Methylquinoline
EINECSNumber (5)-0796
RTECSNumber VC0550000
TSCA Yes