missing translation for 'onlineSavingsMsg'
Learn More

6-Methylquinoline, 98%, Thermo Scientific™

 

Supplier:  Thermo Scientific Chemicals 295140050

Catalog No. AC295140050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

6-Methylquinoline
1.0600g/mL
>113°C
97.5% min. (GC)
1.6125 to 1.6145
5g
quinoline, 6-methyl, p-toluquinoline, 6-methyl-quinoline, tolliquinoline, p, ccris 407, fema no. 2744, quinoline, 6-methyl-8ci,9ci, dsstox_cid_887, dsstox_rid_75844, dsstox_gsid_20887
LUYISICIYVKBTA-UHFFFAOYSA-N
6-methylquinoline
7059
98%
91-62-3
256.0°C to 260.0°C
Authentic
C10H9N
MFCD00006804
1.06
Solubility in water: .. Other solubilities: soluble in ethanol
CC1=CC=C2N=CC=CC2=C1
143.19
143.19

Chemical Identifiers

91-62-3
143.19
LUYISICIYVKBTA-UHFFFAOYSA-N
7059
CC1=CC=C2N=CC=CC2=C1
C10H9N
MFCD00006804
quinoline, 6-methyl, p-toluquinoline, 6-methyl-quinoline, tolliquinoline, p, ccris 407, fema no. 2744, quinoline, 6-methyl-8ci,9ci, dsstox_cid_887, dsstox_rid_75844, dsstox_gsid_20887
6-methylquinoline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.

GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before re

GHS Signal Word: Warning

EINECSNumber : 202-084-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only