missing translation for 'onlineSavingsMsg'
Learn More

6-Methylquinoline, 98%

Catalog No. aab2189214
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
5 g
Catalog No. Quantity
AAB2189214 25 g
AAB2189206 5 g
AAB2189222 100 g
3 options

Catalog No. AAB2189214

Supplier: Thermo Scientific Chemicals B2189214

Only null left

CAS: 91-62-3 | C10H9N | 143.19 g/mol

6-Methylquinoline can be used as primary carbon source in culture of Pseudomonas putida QP1. 6-Methylquinoline was used in the synthesis of fluorescent halide-sensitive quinolinium dyes and fluorescent probes for determination of chloride in biological systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methylquinoline can be used as primary carbon source in culture of Pseudomonas putida QP1. 6-Methylquinoline was used in the synthesis of fluorescent halide-sensitive quinolinium dyes and fluorescent probes for determination of chloride in biological systems.

Solubility
Not miscible or difficult to mix with water.

Notes
Light Sensitive, store in dark. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 91-62-3
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.19
MDL Number MFCD00006804
InChI Key LUYISICIYVKBTA-UHFFFAOYSA-N
Synonym quinoline, 6-methyl, p-toluquinoline, 6-methyl-quinoline, tolliquinoline, p, ccris 407, fema no. 2744, quinoline, 6-methyl-8ci,9ci, dsstox_cid_887, dsstox_rid_75844, dsstox_gsid_20887
PubChem CID 7059
IUPAC Name 6-methylquinoline
SMILES CC1=CC=C2N=CC=CC2=C1

Specifications

Melting Point -22°C
Density 1.066
Boiling Point 257°C to 259°C
Flash Point >110°C (230°F)
Refractive Index 1.615
Quantity 25 g
Beilstein 110336
Sensitivity Light sensitive
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 143.19
Percent Purity 98%
Chemical Name or Material 6-Methylquinoline
Show More Show Less
Hazard Category H302-H315
Hazard Statement GHS H Statement
H301-H341-H315-H319
Toxic if swallowed.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
EINECSNumber 202-084-6
RTECSNumber VC0550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only