missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Methylpyridine-2-carboxylic Acid 98.0+%, TCI America™
$85.75 - $283.33
Chemical Identifiers
| CAS | 934-60-1 |
|---|---|
| Molecular Formula | C7H7NO2 |
| Molecular Weight (g/mol) | 137.14 |
| MDL Number | MFCD00023481 |
| InChI Key | LTUUGSGSUZRPRV-UHFFFAOYSA-N |
| Synonym | 6-methyl-2-pyridinecarboxylic acid, 6-methylpicolinic acid, 6-methyl-2-picolinic acid, 2-carboxy-6-methylpyridine, 6-methyl-2-picolinicacid, 2-pyridinecarboxylic acid, 6-methyl, 2-picoline-6-carboxylic acid, picolinic acid, 6-methyl, 6-methyl picolinic acid, unii-kb675y352l |
| PubChem CID | 70282 |
| IUPAC Name | 6-methylpyridine-2-carboxylic acid |
| SMILES | CC1=CC=CC(=N1)C(O)=O |
Chemical Identifiers
| 934-60-1 | |
| 137.14 | |
| LTUUGSGSUZRPRV-UHFFFAOYSA-N | |
| 70282 | |
| CC1=CC=CC(=N1)C(O)=O |
| C7H7NO2 | |
| MFCD00023481 | |
| 6-methyl-2-pyridinecarboxylic acid, 6-methylpicolinic acid, 6-methyl-2-picolinic acid, 2-carboxy-6-methylpyridine, 6-methyl-2-picolinicacid, 2-pyridinecarboxylic acid, 6-methyl, 2-picoline-6-carboxylic acid, picolinic acid, 6-methyl, 6-methyl picolinic acid, unii-kb675y352l | |
| 6-methylpyridine-2-carboxylic acid |
Specifications
| 934-60-1 | |
| White-Yellow | |
| MFCD00023481 | |
| 6-methyl-2-pyridinecarboxylic acid, 6-methylpicolinic acid, 6-methyl-2-picolinic acid, 2-carboxy-6-methylpyridine, 6-methyl-2-picolinicacid, 2-pyridinecarboxylic acid, 6-methyl, 2-picoline-6-carboxylic acid, picolinic acid, 6-methyl, 6-methyl picolinic acid, unii-kb675y352l | |
| CC1=CC=CC(=N1)C(O)=O | |
| 137.14 | |
| 137.14 | |
| Crystalline Powder |
| 130°C | |
| C7H7NO2 | |
| 5 g | |
| LTUUGSGSUZRPRV-UHFFFAOYSA-N | |
| 6-methylpyridine-2-carboxylic acid | |
| 70282 | |
| ≥98.0% (HPLC,T) | |
| 6-Methylpyridine-2-carboxylic Acid |
Safety and Handling
TSCA : No