missing translation for 'onlineSavingsMsg'
Learn More

6-Methylpyridine-2-carboxaldehyde, 98%

Catalog No. AAH2684106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2684106 5 g
AAH2684114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2684106 Supplier Thermo Scientific Chemicals Supplier No. H2684106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1122-72-1 | C7H7NO | 121.139 g/mol

6-Methyl-2-pyridinecarboxaldehyde was used in the synthesis of 1-(10-cyano- and -10-bromo-5H-dibenzo[a,d]cyclohepten-5-yl)-4-[(arylmethylene)amino]piperazines which are compounds that exhibits anticon vulsant properties. 6-Methylpyridine-2-carboxaldehyde is used to produce 1-(6-methyl-pyridin-2-yl)-hexan-1-one with pent-1-ene at temperature of 100°C. This reaction will need reagent 2-amino-3-picoline and catalysts (Ph3P)3RhCl, Cp2ZrCl2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methyl-2-pyridinecarboxaldehyde was used in the synthesis of 1-(10-cyano- and -10-bromo-5H-dibenzo[a,d]cyclohepten-5-yl)-4-[(arylmethylene)amino]piperazines which are compounds that exhibits anticon vulsant properties. 6-Methylpyridine-2-carboxaldehyde is used to produce 1-(6-methyl-pyridin-2-yl)-hexan-1-one with pent-1-ene at temperature of 100°C. This reaction will need reagent 2-amino-3-picoline and catalysts (Ph3P)3RhCl, Cp2ZrCl2.

Solubility
Soluble in water (750 g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 1122-72-1
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.139
MDL Number MFCD00006291
InChI Key AHISYUZBWDSPQL-UHFFFAOYSA-N
Synonym 6-methyl-2-pyridinecarboxaldehyde, 6-methylpicolinaldehyde, 2-formyl-6-methylpyridine, 6-methylpyridine-2-carboxaldehyde, 2-pyridinecarboxaldehyde, 6-methyl, 6-methylpyridine-2-aldehyde, 2-formyl-6-picoline, 6-methyl-2-pyridinecarbaldehyde, 6-methyl-2-picolinaldehyde, 6-methylpyridyl-2-aldehyde
PubChem CID 70737
IUPAC Name 6-methylpyridine-2-carbaldehyde
SMILES CC1=CC=CC(=N1)C=O

Specifications

Melting Point 29°C to 33°C
Boiling Point 105°C to 108°C (12 mmHg)
Flash Point 68°C (155°F)
Refractive Index 1.527
Quantity 5 g
Beilstein 107476
Solubility Information Soluble in water (750g/L).
Formula Weight 121.14
Percent Purity 98%
Chemical Name or Material 6-Methylpyridine-2-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-359-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only