Learn More
6-Methylphthalazine 98.0+%, TCI America™
Supplier:  TCI America M13941G
Catalog No. M13941G
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
6-Methylphthalazine
White
MFCD03844800
NPOVJYAUMRFAQD-UHFFFAOYSA-N
6-methylphthalazine
12679508
≥98.0% (GC,T)
78032-05-0
C9H8N2
1 g
CC1=CC=C2C=NN=CC2=C1
144.18
144.18
Crystalline Powder
Chemical Identifiers
78032-05-0
144.18
NPOVJYAUMRFAQD-UHFFFAOYSA-N
6-methylphthalazine
C9H8N2
MFCD03844800
12679508
CC1=CC=C2C=NN=CC2=C1
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
TSCA : No