missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Methylnicotinic Acid 98.0+%, TCI America™
$149.13 - $592.76
Chemical Identifiers
| CAS | 3222-47-7 |
|---|---|
| Molecular Formula | C7H7NO2 |
| Molecular Weight (g/mol) | 137.138 |
| MDL Number | MFCD00006341 |
| InChI Key | RZOKQIPOABEQAM-UHFFFAOYSA-N |
| Synonym | 6-methylnicotinic acid, 3-pyridinecarboxylic acid, 6-methyl, 6-methyl nicotinic acid, 2-methylpyridine-5-carboxylic acid, 2-methyl-5-pyridinecarboxylic acid, 6-methyl-3-pyridinecarboxylic acid, 2-methyl-5-pyridine carboxylic acid, 6-methylnicotinate, 6-methylnicotinicacid, pubchem1218 |
| PubChem CID | 137860 |
| IUPAC Name | 6-methylpyridine-3-carboxylic acid |
| SMILES | CC1=NC=C(C=C1)C(=O)O |
Chemical Identifiers
| 3222-47-7 | |
| 137.138 | |
| RZOKQIPOABEQAM-UHFFFAOYSA-N | |
| 137860 | |
| CC1=NC=C(C=C1)C(=O)O |
| C7H7NO2 | |
| MFCD00006341 | |
| 6-methylnicotinic acid, 3-pyridinecarboxylic acid, 6-methyl, 6-methyl nicotinic acid, 2-methylpyridine-5-carboxylic acid, 2-methyl-5-pyridinecarboxylic acid, 6-methyl-3-pyridinecarboxylic acid, 2-methyl-5-pyridine carboxylic acid, 6-methylnicotinate, 6-methylnicotinicacid, pubchem1218 | |
| 6-methylpyridine-3-carboxylic acid |
Specifications
| 3222-47-7 | |
| C7H7NO2 | |
| 5 g | |
| RZOKQIPOABEQAM-UHFFFAOYSA-N | |
| 6-methylpyridine-3-carboxylic acid | |
| 137860 | |
| ≥98.0% (GC,T) | |
| 6-Methylnicotinic Acid |
| 210°C | |
| MFCD00006341 | |
| 6-methylnicotinic acid, 3-pyridinecarboxylic acid, 6-methyl, 6-methyl nicotinic acid, 2-methylpyridine-5-carboxylic acid, 2-methyl-5-pyridinecarboxylic acid, 6-methyl-3-pyridinecarboxylic acid, 2-methyl-5-pyridine carboxylic acid, 6-methylnicotinate, 6-methylnicotinicacid, pubchem1218 | |
| CC1=NC=C(C=C1)C(=O)O | |
| 137.138 | |
| 137.14 | |
| Crystalline Powder |
Safety and Handling
TSCA : No