missing translation for 'onlineSavingsMsg'
Learn More

6-Methylnicotinic acid, 99%

Catalog No. AC307620050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC307620050 5 g Glass bottle
AC307620010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC307620050 Supplier Thermo Scientific Chemicals Supplier No. 307620050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3222-47-7 | C7H7NO2 | 137.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3222-47-7
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.17
MDL Number MFCD00006341
InChI Key RZOKQIPOABEQAM-UHFFFAOYSA-N
Synonym 6-methylnicotinic acid, 3-pyridinecarboxylic acid, 6-methyl, 6-methyl nicotinic acid, 2-methylpyridine-5-carboxylic acid, 2-methyl-5-pyridinecarboxylic acid, 6-methyl-3-pyridinecarboxylic acid, 2-methyl-5-pyridine carboxylic acid, 6-methylnicotinate, 6-methylnicotinicacid, pubchem1218
PubChem CID 137860
IUPAC Name 6-methylpyridine-3-carboxylic acid
SMILES CC1=NC=C(C=C1)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 209°C to 213°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Formula Weight 137.17
Percent Purity 99%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material 6-Methylnicotinic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only