Learn More
6-Methylindole, 98%
CAS: 3420-02-8 | C9H9N | 131.18 g/mol
Supplier: Thermo Scientific Chemicals L17509MD
Description
It is a reactant for preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators, reactant for preparation of indole linked triazole derivatives as antifungal agents, reactant for preparation of N-β-D-xylosyl-indole derivatives as SGLT2 inhibitors for management of hyperglycemia in diabetes, reactant for preparation of aminoguanidine derivatives of arylsulfonylacylindoles as antifungal agents, reactant for preparation of indolylindazoles and indolylpyrazolopyridines as interleukin-2 inducible T cell kinase inhibitors, reactant for preparation of arylsulfonylacetylindoles as anti-HIV-1 agents. 6-Methylindole was used in the synthesis of benz[c,d]indol-3(1H)-one derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a reactant for preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators, reactant for preparation of indole linked triazole derivatives as antifungal agents, reactant for preparation of N-β-D-xylosyl-indole derivatives as SGLT2 inhibitors for management of hyperglycemia in diabetes, reactant for preparation of aminoguanidine derivatives of arylsulfonylacylindoles as antifungal agents, reactant for preparation of indolylindazoles and indolylpyrazolopyridines as interleukin-2 inducible T cell kinase inhibitors, reactant for preparation of arylsulfonylacetylindoles as anti-HIV-1 agents. 6-Methylindole was used in the synthesis of benz[c,d]indol-3(1H)-one derivatives.
Solubility
Insoluble in water.
Notes
Moisture and light sensitive. Store in dry condition. Keep away from active metals, bases, oxidizing agents and heat.
Specifications
| 6-Methylindole | |
| 29°C to 32°C | |
| 112°C (5 mmHg) | |
| C11H15NO2 | |
| MFCD00005682 | |
| 109708 | |
| n-boc-aniline, tert-butyl phenylcarbamate, n-boc aniline, t-butyl carbanilate, tert-butyl carbanilate, carbanilic acid, tert-butyl ester, carbamic acid, phenyl-, 1,1-dimethylethyl ester, n-tert-butoxycarbonyl aniline, tert-butyl n-phenylurethane, carbanilic acid tert-butyl ester | |
| KZZHPWMVEVZEFG-UHFFFAOYSA-N | |
| tert-butyl N-phenylcarbamate | |
| 137930 | |
| 98% |
| 3420-02-8 | |
| 1.059 | |
| >110°C (230°F) | |
| 1.6071 | |
| 250 mg | |
| Light sensitive | |
| Insoluble in water. | |
| CC(C)(C)OC(=O)NC1=CC=CC=C1 | |
| 193.25 | |
| 131.18 |
Chemical Identifiers
| 3420-02-8 | |
| MFCD00005682 |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only