missing translation for 'onlineSavingsMsg'
Learn More

6-Methylheptane-2,4-dione, 98+%

Catalog No. p-7021151
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1337304 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1337304 Supplier Thermo Scientific Chemicals Supplier No. A1337304

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3002-23-1 | C8H14O2 | 142.198 g/mol

6-Methylheptane-2,4-dione, is used as a multipurpose intermediate. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceutical industry

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methylheptane-2,4-dione, is used as a multipurpose intermediate. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceutical industry

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from Strong oxidizing agents.

Chemical Identifiers

CAS 3002-23-1
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00008939
InChI Key IGMOYJSFRIASIE-UHFFFAOYSA-N
Synonym 6-methyl-2,4-heptanedione, isovalerylacetone, 2,4-heptanedione, 6-methyl, unii-o97vug08ub, 6-methylheptan-2,4-dione, o97vug08ub, nsc94, nsc 94, acmc-1ct2p, 6-methyl-2,4-heptandione
PubChem CID 76354
IUPAC Name 6-methylheptane-2,4-dione
SMILES CC(C)CC(=O)CC(=O)C

Specifications

Density 0.919
Boiling Point 183°C
Flash Point 58°C (136°F)
Refractive Index 1.456
Quantity 2 g
UN Number UN1224
Beilstein 1751122
Solubility Information It is insoluble in water.
Formula Weight 142.2
Percent Purity ≥98%
Chemical Name or Material 6-Methylheptane-2,4-dione
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 221-093-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only