missing translation for 'onlineSavingsMsg'
Learn More

6-Methylflavone 98.0+%, TCI America™

Catalog No. M27665G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27665G 5 g
M27661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27665G Supplier TCI America Supplier No. M27665G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 29976-75-8
Molecular Formula C16H12O2
Molecular Weight (g/mol) 236.27
MDL Number MFCD00017461
InChI Key NOQJBXPAMJLUSS-UHFFFAOYSA-N
PubChem CID 689013
IUPAC Name 6-methyl-2-phenylchromen-4-one
SMILES CC1=CC2=C(C=C1)OC(=CC2=O)C3=CC=CC=C3

Specifications

Melting Point 121°C
Color White-Yellow
Boiling Point 235°C
Quantity 5 g
Formula Weight 236.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methylflavone