missing translation for 'onlineSavingsMsg'
Learn More

6-Methylcoumarin 99.0+%, TCI America™

Supplier:  TCI America M018925G

Catalog No. M018925G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

6-Methylcoumarin
76°C
200°C
MFCD00006875
6-methylcoumarin, 6-methylbenzopyrone, 6-methyl-2h-chromen-2-one, toncarine, 6-methyl coumarin, methyl coumarin, 6-methylcoumarinic anhydride, coumarin, 6-methyl, 6-methyl-1,2-benzopyrone, 6-methylcumarin
CC1=CC=C2OC(=O)C=CC2=C1
160.17
CHEBI:563586
≥99.0% (GC)
92-48-8
White-Yellow
C10H8O2
25 g
FXFYOPQLGGEACP-UHFFFAOYSA-N
6-methyl-2H-chromen-2-one
7092
160.17
Crystalline Powder

Chemical Identifiers

92-48-8
160.17
FXFYOPQLGGEACP-UHFFFAOYSA-N
7092
6-methyl-2H-chromen-2-one
C10H8O2
MFCD00006875
6-methylcoumarin, 6-methylbenzopyrone, 6-methyl-2h-chromen-2-one, toncarine, 6-methyl coumarin, methyl coumarin, 6-methylcoumarinic anhydride, coumarin, 6-methyl, 6-methyl-1,2-benzopyrone, 6-methylcumarin
CHEBI:563586
CC1=CC=C2OC(=O)C=CC2=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0687

RTECSNumber : GN7792000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions