missing translation for 'onlineSavingsMsg'
Learn More

6-Methylcoumarin, 99%

Catalog No. AAA1938218
Change view
Click to view available options
Quantity:
50 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1938218 50 g
AAA1938222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1938218 Supplier Thermo Scientific Chemicals Supplier No. A1938218
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 92-48-8 | C10H8O2 | 160.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 92-48-8
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.17
MDL Number MFCD00006875
InChI Key FXFYOPQLGGEACP-UHFFFAOYSA-N
Synonym 6-methylcoumarin, 6-methylbenzopyrone, 6-methyl-2h-chromen-2-one, toncarine, 6-methyl coumarin, methyl coumarin, 6-methylcoumarinic anhydride, coumarin, 6-methyl, 6-methyl-1,2-benzopyrone, 6-methylcumarin
PubChem CID 7092
ChEBI CHEBI:563586
IUPAC Name 6-methylchromen-2-one
SMILES CC1=CC=C2OC(=O)C=CC2=C1

Specifications

Melting Point 74°C to 77°C
Boiling Point 303°C (725 mmHg)
Quantity 50 g
Beilstein 4222
Formula Weight 160.17
Percent Purity 99%
Chemical Name or Material 6-Methylcoumarin
Hazard Category H301+H311+H331
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c
EINECSNumber 202-158-8
RTECSNumber GN7792000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only