missing translation for 'onlineSavingsMsg'
Learn More

6-Methylchromone 98.0+%, TCI America™

Catalog No. p-7133414
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
M27641G 1 g
M27645G 5 g
2 options

Catalog No. M27641G

Supplier: TCI America M27641G

Only null left

Chemical Identifiers

CAS 38445-23-7
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.172
MDL Number MFCD00218598
InChI Key HTXQVFXXVXOLCF-UHFFFAOYSA-N
Synonym 6-methylchromone, 6-methyl-4h-chromen-4-one, 6-methylchromone hydrate, chromone, 6-methyl, acmc-20aly0, 6-methyl-4h-chromen-4-one #, 6-methyl-4-oxo-4h-1-benzopyran, 6-methyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one,6-methyl
PubChem CID 594810
IUPAC Name 6-methylchromen-4-one
SMILES CC1=CC2=C(C=C1)OC=CC2=O

Specifications

Melting Point 87°C
Color Yellow
Quantity 1 g
Formula Weight 160.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methylchromone