missing translation for 'onlineSavingsMsg'
Learn More

6-Methylchromone, 98%

Catalog No. AAH2700203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2700203 1 g
AAH2700206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2700203 Supplier Thermo Scientific Chemicals Supplier No. H2700203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 38445-23-7 | C10H8O2 | 160.172 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 38445-23-7
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.172
MDL Number MFCD00218598
InChI Key HTXQVFXXVXOLCF-UHFFFAOYSA-N
Synonym 6-methylchromone, 6-methyl-4h-chromen-4-one, 6-methylchromone hydrate, chromone, 6-methyl, acmc-20aly0, 6-methyl-4h-chromen-4-one #, 6-methyl-4-oxo-4h-1-benzopyran, 6-methyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one,6-methyl
PubChem CID 594810
IUPAC Name 6-methylchromen-4-one
SMILES CC1=CC2=C(C=C1)OC=CC2=O

Specifications

Melting Point 88°C to 90°C
Quantity 1 g
Formula Weight 160.17
Percent Purity 98%
Chemical Name or Material 6-Methylchromone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only