missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-5-propyl-4(1H)-pyrimidinone 98.0+%, TCI America™

Catalog No. M16355G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M16355G Supplier TCI America Supplier No. M16355G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 103980-68-3
Molecular Formula C8H12N2O
Molecular Weight (g/mol) 152.197
MDL Number MFCD01689111
InChI Key UAJIFHDSCLQUON-UHFFFAOYSA-N
PubChem CID 3025555
IUPAC Name 6-methyl-5-propyl-1H-pyrimidin-4-one
SMILES CCCC1=C(NC=NC1=O)C

Specifications

Melting Point 116°C
Color White
Quantity 5 g
Formula Weight 152.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methyl-5-propyl-4(1H)-pyrimidinone
RTECSNumber UW6980000
TSCA No