missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-5-hepten-2-one 98.0+%, TCI America™

Catalog No. M025225ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M025225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M025225ML Supplier TCI America Supplier No. M025225ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-93-0
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.20
MDL Number MFCD00008905
InChI Key UHEPJGULSIKKTP-UHFFFAOYSA-N
Synonym 6-methyl-5-hepten-2-one, sulcatone, methylheptenone, 5-hepten-2-one, 6-methyl, 2-methyl-2-hepten-6-one, methyl heptenone, 6-methyl-5-heptene-2-one, heptenone, methyl, 2-methyl-6-oxo-2-heptene, 2-oxo-6-methylhept-5-ene
PubChem CID 9862
ChEBI CHEBI:16310
IUPAC Name 6-methylhept-5-en-2-one
SMILES CC(C)=CCCC(C)=O

Specifications

Color Yellow
Boiling Point 173°C
Quantity 25 mL
UN Number 1224
Formula Weight 126.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 6-Methyl-5-hepten-2-one
EINECSNumber (2)-2480
RTECSNumber MJ9700000
TSCA Yes