missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-5-hepten-2-one, 98%

Catalog No. AAA1333522
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1333522 100 g
AAA1333514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1333522 Supplier Thermo Scientific Chemicals Supplier No. A1333522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 110-93-0 | C8H14O | 126.20 g/mol

6-Methyl-hepten-2-one is used in the synthesis of thyrsiferyl 23-Acetate which acts as an anti-leukemic inducer of apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methyl-hepten-2-one is used in the synthesis of thyrsiferyl 23-Acetate which acts as an anti-leukemic inducer of apoptosis.

Solubility
Soluble in methanol and chloroform.

Notes
Store in cool, dry place in tightly closed container.

Chemical Identifiers

CAS 110-93-0
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.20
MDL Number MFCD00008905
InChI Key UHEPJGULSIKKTP-UHFFFAOYSA-N
Synonym 6-methyl-5-hepten-2-one, sulcatone, methylheptenone, 5-hepten-2-one, 6-methyl, 2-methyl-2-hepten-6-one, methyl heptenone, 6-methyl-5-heptene-2-one, heptenone, methyl, 2-methyl-6-oxo-2-heptene, 2-oxo-6-methylhept-5-ene
PubChem CID 9862
ChEBI CHEBI:16310
IUPAC Name 6-methylhept-5-en-2-one
SMILES CC(C)=CCCC(C)=O

Specifications

Melting Point -67°C
Density 0.855
Boiling Point 172°C to 174°C
Flash Point 56°C (133°F)
Refractive Index 1.439
Quantity 100 g
UN Number UN1224
Beilstein 1741705
Solubility Information Soluble in methanol and chloroform.
Formula Weight 126.2
Percent Purity 98%
Chemical Name or Material 6-Methyl-5-hepten-2-one
Show More Show Less
Hazard Category H226-H319
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 203-816-7
RTECSNumber MJ9700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only