missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-6-Methyl-5-hepten-2-ol, 98%

Catalog No. aal0918114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0918114 25 g
AAL0918106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0918114 Supplier Thermo Scientific Chemicals Supplier No. L0918114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1569-60-4 | C8H16O | 128.215 g/mol

(±)-6-Methyl-5-hepten-2-ol is a compound used in the synthesis of aggregation pheromone of the ambrosia beetle. It is also used as pharmaceutical intermediates and insect attractant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+/-)-6-Methyl-5-hepten-2-ol is a compound used in the synthesis of aggregation pheromone of the ambrosia beetle. It is also used as pharmaceutical intermediates and insect attractant.

Solubility
Soluble in Chloroform. Not miscible with water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 1569-60-4
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00004561
InChI Key OHEFFKYYKJVVOX-UHFFFAOYSA-N
Synonym 6-methyl-5-hepten-2-ol, sulcatol, 5-hepten-2-ol, 6-methyl, r-sulcatol, 2-methyl-2-hepten-6-ol, +/--6-methyl-5-hepten-2-ol, 6-methyl-hept-5-en-2-ol, r---6-methyl-5-hepten-2-ol, 6-hydroxy-2-methyl-2-heptene, acmc-20apkw
PubChem CID 20745
ChEBI CHEBI:15833
IUPAC Name 6-methylhept-5-en-2-ol
SMILES CC(CCC=C(C)C)O

Specifications

Density 0.84
Boiling Point 78°C to 80°C (15 mmHg)
Flash Point 67°C (152°F)
Refractive Index 1.448
Quantity 25 g
Beilstein 1720073
Solubility Information Soluble in Chloroform. Not miscible with water.
Formula Weight 128.22
Percent Purity 98%
Chemical Name or Material (+/-)-6-Methyl-5-hepten-2-ol
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 216-377-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only