missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Methyl-3,5-heptadiene-2-one, 96%, Thermo Scientific™
$142.27 - $142.27
Chemical Identifiers
| CAS | 1604-28-0 |
|---|---|
| Molecular Formula | C8H12O |
| Molecular Weight (g/mol) | 124.18 |
| InChI Key | KSKXSFZGARKWOW-GQCTYLIASA-N |
| Synonym | 6-methyl-3,5-heptadien-2-one, 6-methylhepta-3,5-dien-2-one, methylheptadienone, unii-p7cmp2e76c, 3,5-heptadien-2-one, 6-methyl, 3e-6-methylhepta-3,5-dien-2-one, 3,5-heptadien-2-one, 6-methyl-, e, 6-methyl-trans-3,5-heptadien-2-one, p7cmp2e76c, fema no. 3363 |
| PubChem CID | 5370101 |
| IUPAC Name | (3E)-6-methylhepta-3,5-dien-2-one |
| SMILES | CC(=CC=CC(=O)C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC453210250
|
Thermo Scientific Chemicals
453210250 |
25 g |
N/A
|
|
|||||
Chemical Identifiers
| 1604-28-0 | |
| 124.18 | |
| 6-methyl-3,5-heptadien-2-one, 6-methylhepta-3,5-dien-2-one, methylheptadienone, unii-p7cmp2e76c, 3,5-heptadien-2-one, 6-methyl, 3e-6-methylhepta-3,5-dien-2-one, 3,5-heptadien-2-one, 6-methyl-, e, 6-methyl-trans-3,5-heptadien-2-one, p7cmp2e76c, fema no. 3363 | |
| (3E)-6-methylhepta-3,5-dien-2-one |
| C8H12O | |
| KSKXSFZGARKWOW-GQCTYLIASA-N | |
| 5370101 | |
| CC(=CC=CC(=O)C)C |
Specifications
| 1604-28-0 | |
| 168.0°C | |
| Authentic | |
| C8H12O | |
| 25 g | |
| 6-methyl-3,5-heptadien-2-one, 6-methylhepta-3,5-dien-2-one, methylheptadienone, unii-p7cmp2e76c, 3,5-heptadien-2-one, 6-methyl, 3e-6-methylhepta-3,5-dien-2-one, 3,5-heptadien-2-one, 6-methyl-, e, 6-methyl-trans-3,5-heptadien-2-one, p7cmp2e76c, fema no. 3363 | |
| CC(=CC=CC(=O)C)C | |
| 124.18 | |
| 124.18 | |
| 6-Methyl-3,5-heptadiene-2-one |
| 0.8930g/mL | |
| 80°C | |
| 95% min. (GC) | |
| (CH3)2C=CHCH=CHCOCH3 | |
| 0.893 | |
| KSKXSFZGARKWOW-GQCTYLIASA-N | |
| (3E)-6-methylhepta-3,5-dien-2-one | |
| 5370101 | |
| 96% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 216-507-7
RUO – Research Use Only