missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-3,5-heptadiene-2-one, 96%, Thermo Scientific™

Catalog No. AC453210250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453210250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC453210250 Supplier Thermo Scientific Chemicals Supplier No. 453210250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1604-28-0
Molecular Formula C8H12O
Molecular Weight (g/mol) 124.18
InChI Key KSKXSFZGARKWOW-GQCTYLIASA-N
Synonym 6-methyl-3,5-heptadien-2-one, 6-methylhepta-3,5-dien-2-one, methylheptadienone, unii-p7cmp2e76c, 3,5-heptadien-2-one, 6-methyl, 3e-6-methylhepta-3,5-dien-2-one, 3,5-heptadien-2-one, 6-methyl-, e, 6-methyl-trans-3,5-heptadien-2-one, p7cmp2e76c, fema no. 3363
PubChem CID 5370101
IUPAC Name (3E)-6-methylhepta-3,5-dien-2-one
SMILES CC(=CC=CC(=O)C)C

Specifications

Density 0.8930g/mL
Boiling Point 168.0°C
Flash Point 80°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Linear Formula (CH3)2C=CHCH=CHCOCH3
Quantity 25 g
Specific Gravity 0.893
Formula Weight 124.18
Percent Purity 96%
Chemical Name or Material 6-Methyl-3,5-heptadiene-2-one
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 216-507-7

RUO – Research Use Only