missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-3,5-heptadien-2-one, 97%

Catalog No. p-7045027
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0916906 5 g
AAL0916914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0916906 Supplier Thermo Scientific Chemicals Supplier No. L0916906
Only null left

CAS: 1604-28-0 | C8H12O | 124.183 g/mol

6-Methyl-3,5-heptadien-2-one is used in the preparation of (RS)-(E)-2,6-dimethyl-1,2-epoxy-3,5-heptadiene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methyl-3,5-heptadien-2-one is used in the preparation of (RS)-(E)-2,6-dimethyl-1,2-epoxy-3,5-heptadiene.

Solubility
Miscible with alcohol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 1604-28-0
Molecular Formula C8H12O
Molecular Weight (g/mol) 124.183
MDL Number MFCD00043647
InChI Key KSKXSFZGARKWOW-GQCTYLIASA-N
Synonym 6-methyl-3,5-heptadien-2-one, 6-methylhepta-3,5-dien-2-one, methylheptadienone, unii-p7cmp2e76c, 3,5-heptadien-2-one, 6-methyl, 3e-6-methylhepta-3,5-dien-2-one, 3,5-heptadien-2-one, 6-methyl-, e, 6-methyl-trans-3,5-heptadien-2-one, p7cmp2e76c, fema no. 3363
PubChem CID 5370101
IUPAC Name (3E)-6-methylhepta-3,5-dien-2-one
SMILES CC(=CC=CC(=O)C)C

Specifications

Density 0.89
Boiling Point 95°C to 96°C (20 mmHg)
Odor Pleasant
Refractive Index 1.534
Quantity 5 g
Beilstein 1699937
Solubility Information Miscible with alcohol. Immiscible with water.
Formula Weight 124.18
Percent Purity 97%
Chemical Name or Material 6-Methyl-3,5-heptadien-2-one
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 216-507-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only