missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-3(2H)-pyridazinone 98.0+%, TCI America™

Catalog No. p-7133172
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
M23441G 1 g
M23445G 5 g
2 options

Catalog No. M23441G

Supplier: TCI America M23441G

Only null left

Chemical Identifiers

CAS 13327-27-0
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD00039720
InChI Key QZWIXLPWMGHDDD-UHFFFAOYSA-N
Synonym 6-methylpyridazin-3 2h-one, 6-methyl-3 2h-pyridazinone, 6-methylpyridazin-3-ol, 6-methyl-2h-pyridazin-3-one, 6-methyl-3-pyridazinone, 3-hydroxy-6-methylpyridazine, 3 2h-pyridazinone, 6-methyl, 6-methyl-2,3-dihydropyridazin-3-one, 6me3oxopyridaz
PubChem CID 83346
IUPAC Name 6-methyl-2,3-dihydropyridazin-3-one
SMILES CC1=NNC(=O)C=C1

Specifications

Melting Point 144°C
Color Yellow
Quantity 1 g
Formula Weight 110.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methyl-3(2H)-pyridazinone