missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-3(2H)-pyridazinone, 98%

Catalog No. AAA1447106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1447106 5 g
AAA1447114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1447106 Supplier Thermo Scientific Chemicals Supplier No. A1447106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13327-27-0 | C5H6N2O | 110.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13327-27-0
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD00039720
InChI Key QZWIXLPWMGHDDD-UHFFFAOYSA-N
Synonym 6-methylpyridazin-3 2h-one, 6-methyl-3 2h-pyridazinone, 6-methylpyridazin-3-ol, 6-methyl-2h-pyridazin-3-one, 6-methyl-3-pyridazinone, 3-hydroxy-6-methylpyridazine, 3 2h-pyridazinone, 6-methyl, 6-methyl-2,3-dihydropyridazin-3-one, 6me3oxopyridaz
PubChem CID 83346
IUPAC Name 3-methyl-1H-pyridazin-6-one
SMILES CC1=NNC(=O)C=C1

Specifications

Melting Point 143°C to 145°C
Quantity 5 g
Beilstein 109928
Formula Weight 110.12
Percent Purity 98%
Chemical Name or Material 6-Methyl-3(2H)-pyridazinone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 236-367-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only