missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-2-pyridinemethanol 98.0+%, TCI America™

Catalog No. p-7132642
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12825G 5 g
M128225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12825G Supplier TCI America Supplier No. M12825G
Only null left

Chemical Identifiers

CAS 1122-71-0
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.16
MDL Number MFCD00023520
InChI Key JLVBSBMJQUMAMW-UHFFFAOYSA-N
Synonym 6-methyl-2-pyridinemethanol, 6-methylpyridin-2-yl methanol, 2-pyridinemethanol, 6-methyl, 6-methyl-2-pyridylmethanol, 6-methylpyridine-2-methanol, 2-hydroxymethyl-6-methylpyridine, 6-methyl-2-pyridyl methan-1-ol, pubchem20959, acmc-2099dw, 6-methyl-2-pyridylcarbinol
PubChem CID 70736
IUPAC Name (6-methylpyridin-2-yl)methanol
SMILES CC1=CC=CC(CO)=N1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 123.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 6-Methyl-2-pyridinemethanol