missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-2-pyridinemethanol, 98%

Catalog No. AAH2679414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2679414 25 g
AAH2679406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2679414 Supplier Thermo Scientific Chemicals Supplier No. H2679414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1122-71-0 | C7H9NO | 123.16 g/mol

Use as primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Use as primary and secondary intermediate.

Solubility
Low soluble in water, high soluble in ethanol, acetic acid. Soluble in benzene, toluene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, strong reducing agents. Store under an inert atmosphere.

Chemical Identifiers

CAS 1122-71-0
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.16
MDL Number MFCD00023520
InChI Key JLVBSBMJQUMAMW-UHFFFAOYSA-N
Synonym 6-methyl-2-pyridinemethanol, 6-methylpyridin-2-yl methanol, 2-pyridinemethanol, 6-methyl, 6-methyl-2-pyridylmethanol, 6-methylpyridine-2-methanol, 2-hydroxymethyl-6-methylpyridine, 6-methyl-2-pyridyl methan-1-ol, pubchem20959, acmc-2099dw, 6-methyl-2-pyridylcarbinol
PubChem CID 70736
IUPAC Name (6-methylpyridin-2-yl)methanol
SMILES CC1=CC=CC(CO)=N1

Specifications

Melting Point 32°C to 34°C
Boiling Point 105°C to 108°C (12 mmHg)
Flash Point 109°C (228°F)
Quantity 25 g
Solubility Information Low soluble in water,high soluble in ethanol,acetic acid. Soluble in benzene,toluene.
Formula Weight 123.15
Percent Purity 98%
Chemical Name or Material 6-Methyl-2-pyridinemethanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-358-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only