missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-2-pyridinecarboxaldehyde 98.0+%, TCI America™

Catalog No. M11575G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11575G 5 g
M115725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11575G Supplier TCI America Supplier No. M11575G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1122-72-1
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.139
MDL Number MFCD00006291
InChI Key AHISYUZBWDSPQL-UHFFFAOYSA-N
Synonym 6-methyl-2-pyridinecarboxaldehyde, 6-methylpicolinaldehyde, 2-formyl-6-methylpyridine, 6-methylpyridine-2-carboxaldehyde, 2-pyridinecarboxaldehyde, 6-methyl, 6-methylpyridine-2-aldehyde, 2-formyl-6-picoline, 6-methyl-2-pyridinecarbaldehyde, 6-methyl-2-picolinaldehyde, 6-methylpyridyl-2-aldehyde
PubChem CID 70737
IUPAC Name 6-methylpyridine-2-carbaldehyde
SMILES CC1=CC=CC(=N1)C=O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 121.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 6-Methyl-2-pyridinecarboxaldehyde
TSCA Yes
Recommended Storage Refrigerator