missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-2-(p-tolylamino)-4H-3,1-benzoxazin-4-one 95.0+%, TCI America™

Catalog No. M257250MG
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M257250MG 50 mg
M257210MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M257250MG Supplier TCI America Supplier No. M257250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 86672-58-4
Molecular Formula C16H14N2O2
Molecular Weight (g/mol) 266.30
MDL Number MFCD03272508
InChI Key GFWNGVKCDGYFKG-UHFFFAOYSA-N
Synonym URB-754, 6-Methyl-2-(p-tolylamino)-4H-benzo[d][1,3]oxazin-4-one
PubChem CID 848487
IUPAC Name 6-methyl-2-[(4-methylphenyl)amino]-4H-3,1-benzoxazin-4-one
SMILES CC1=CC=C(NC2=NC3=CC=C(C)C=C3C(=O)O2)C=C1

Specifications

Melting Point 215°C
Color Yellow
Quantity 50 mg
Formula Weight 266.30
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methyl-2-(p-tolylamino)-4H-3,1-benzoxazin-4-one
TSCA No
Recommended Storage Freezer (-20 °C)