missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-2-heptanol 97.0+%, TCI America™

Catalog No. M07915ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M07915ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M07915ML Supplier TCI America Supplier No. M07915ML

Chemical Identifiers

CAS 4730-22-7
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.231
MDL Number MFCD00004562
InChI Key FCOUHTHQYOMLJT-UHFFFAOYSA-N
PubChem CID 96476
IUPAC Name 6-methylheptan-2-ol
SMILES CC(C)CCCC(C)O

Specifications

Color Yellow
Boiling Point 172°C
Quantity 5 mL
Formula Weight 130.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 6-Methyl-2-heptanol
EINECSNumber (2)-0217
TSCA No