missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-1-indanone 98.0+%, TCI America™

$65.46 - $65.46

Chemical Identifiers

CAS 24623-20-9
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.19
MDL Number MFCD00275729
InChI Key DBOXRDYLMJMQBB-UHFFFAOYSA-N
Synonym 6-methyl-1-indanone, 6-methyl-2,3-dihydro-1h-inden-1-one, 6-methylindan-1-one, 6-methyl-indan-1-one, 1h-inden-1-one, 2,3-dihydro-6-methyl, 2,3-dihydro-6-methylinden-1-one, pubchem8851, acmc-209gdm, 6-methyl-indane-1-one
PubChem CID 2795093
IUPAC Name 6-methyl-2,3-dihydro-1H-inden-1-one
SMILES CC1=CC2=C(CCC2=O)C=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M19015G
View Documents
TCI America
M19015G
5 g
Each for $65.46
Only null left
 
Description

Description

Specifications

Chemical Identifiers

24623-20-9
146.19
DBOXRDYLMJMQBB-UHFFFAOYSA-N
2795093
CC1=CC2=C(CCC2=O)C=C1
C10H10O
MFCD00275729
6-methyl-1-indanone, 6-methyl-2,3-dihydro-1h-inden-1-one, 6-methylindan-1-one, 6-methyl-indan-1-one, 1h-inden-1-one, 2,3-dihydro-6-methyl, 2,3-dihydro-6-methylinden-1-one, pubchem8851, acmc-209gdm, 6-methyl-indane-1-one
6-methyl-2,3-dihydro-1H-inden-1-one

Specifications

24623-20-9
White-Yellow
C10H10O
5 g
DBOXRDYLMJMQBB-UHFFFAOYSA-N
6-methyl-2,3-dihydro-1H-inden-1-one
2795093
≥98.0% (GC)
6-Methyl-1-indanone
62°C
126°C
MFCD00275729
6-methyl-1-indanone, 6-methyl-2,3-dihydro-1h-inden-1-one, 6-methylindan-1-one, 6-methyl-indan-1-one, 1h-inden-1-one, 2,3-dihydro-6-methyl, 2,3-dihydro-6-methylinden-1-one, pubchem8851, acmc-209gdm, 6-methyl-indane-1-one
CC1=CC2=C(CCC2=O)C=C1
146.19
146.19
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'