missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-1-indanone 98.0+%, TCI America™

Catalog No. M19015G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M19015G Supplier TCI America Supplier No. M19015G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 24623-20-9
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.19
MDL Number MFCD00275729
InChI Key DBOXRDYLMJMQBB-UHFFFAOYSA-N
Synonym 6-methyl-1-indanone, 6-methyl-2,3-dihydro-1h-inden-1-one, 6-methylindan-1-one, 6-methyl-indan-1-one, 1h-inden-1-one, 2,3-dihydro-6-methyl, 2,3-dihydro-6-methylinden-1-one, pubchem8851, acmc-209gdm, 6-methyl-indane-1-one
PubChem CID 2795093
IUPAC Name 6-methyl-2,3-dihydro-1H-inden-1-one
SMILES CC1=CC2=C(CCC2=O)C=C1

Specifications

Melting Point 62°C
Color White-Yellow
Boiling Point 126°C
Quantity 5 g
Formula Weight 146.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methyl-1-indanone