missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-1,3-benzoxazole-2(3H)-thione 98.0+%, TCI America™

Catalog No. p-7133558
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M30631G 1 g
M30635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. M30631G missing translation for 'mfr' TCI America missing translation for 'supplierNo' M30631G
Only null left

Chemical Identifiers

CAS 23417-29-0
Molecular Formula C8H7NOS
Molecular Weight (g/mol) 165.21
MDL Number MFCD00087075,MFCD00175644
InChI Key NUMDWMDMUXZMHA-UHFFFAOYSA-N
Synonym 6-Methyl-1,3-benzoxazole-2-thiol
PubChem CID 963501
IUPAC Name 6-methyl-2,3-dihydro-1,3-benzoxazole-2-thione
SMILES CC1=CC=C2NC(=S)OC2=C1

Specifications

Melting Point 211°C
Quantity 1 g
Formula Weight 165.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methyl-1,3-benzoxazole-2(3H)-thione