missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxyquinoline N-Oxide 98.0+%, TCI America™

Catalog No. p-7133527
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29991G 1 g
M29995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M29991G Supplier TCI America Supplier No. M29991G
Only null left

Chemical Identifiers

CAS 6563-13-9
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD00009771
InChI Key BWEGRKPOJXNZSK-UHFFFAOYSA-N
PubChem CID 253821
IUPAC Name 6-methoxy-1-oxidoquinolin-1-ium
SMILES COC1=CC2=C(C=C1)[N+](=CC=C2)[O-]

Specifications

Melting Point 110°C
Quantity 1 g
Formula Weight 175.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methoxyquinoline N-Oxide