missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxyquinoline, 98%

Catalog No. aaa1173614
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1173614 25 g
AAA1173606 5 g
AAA1173622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1173614 Supplier Thermo Scientific Chemicals Supplier No. A1173614
Only null left

CAS: 5263-87-6 | C10H9NO | 159.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5263-87-6
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.188
MDL Number MFCD00006800
InChI Key HFDLDPJYCIEXJP-UHFFFAOYSA-N
Synonym quinoline, 6-methoxy, 6-methoxy-quinoline, methyl 6-quinolyl ether, p-quinanisole, unii-5s1u3125ad, chembl15200, 6-methoxy quinoline, pubchem5917, 6-methoxyquinoline, acmc-209l0q
PubChem CID 14860
ChEBI CHEBI:72822
IUPAC Name 6-methoxyquinoline
SMILES COC1=CC2=C(C=C1)N=CC=C2

Specifications

Melting Point 17°C to 20°C
Density 1.152
Boiling Point 141°C to 145°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.622
Quantity 25 g
Beilstein 115244
Sensitivity Light sensitive
Formula Weight 159.19
Percent Purity 98%
Chemical Name or Material 6-Methoxyquinoline
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 226-077-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only