missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxyquinoline, 98%

Catalog No. AC179341000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC179341000 100 g Glass bottle
AC179340250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC179341000 Supplier Thermo Scientific Chemicals Supplier No. 179341000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5263-87-6 | C10H9NO | 159.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5263-87-6
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.19
MDL Number MFCD00006800
InChI Key HFDLDPJYCIEXJP-UHFFFAOYSA-N
Synonym quinoline, 6-methoxy, 6-methoxy-quinoline, methyl 6-quinolyl ether, p-quinanisole, unii-5s1u3125ad, chembl15200, 6-methoxy quinoline, pubchem5917, 6-methoxyquinoline, acmc-209l0q
PubChem CID 14860
ChEBI CHEBI:72822
IUPAC Name 6-methoxyquinoline
SMILES COC1=CC2=C(C=C1)N=CC=C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 18.0°C to 20.0°C
Color White to Cream
Density 1.1520g/mL
Boiling Point 193.0°C (50.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6170 to 1.6240
Quantity 100 g
Beilstein 21,85
Specific Gravity 1.152
Solubility Information Solubility in water: insoluble
Formula Weight 159.19
Percent Purity 98%
Physical Form Granular Powder
Chemical Name or Material 6-Methoxyquinoline, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not eat,drink or smoke when using this product.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water f
EINECSNumber 226-077-2
TSCA TSCA

RUO – Research Use Only