missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxypyridine-3-carboxaldehyde, 98%

Catalog No. AAH2651503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2651503 1 g
AAH2651506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2651503 Supplier Thermo Scientific Chemicals Supplier No. H2651503

CAS: 65873-72-5 | C7H7NO2 | 137.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 65873-72-5
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD02683446
InChI Key CTAIEPPAOULMFY-UHFFFAOYSA-N
Synonym 6-methoxynicotinaldehyde, 6-methoxy-3-pyridinecarboxaldehyde, 5-formyl-2-methoxypyridine, 2-methoxypyridine-5-carboxaldehyde, 5-formyl-2-methoxy-pyridine, 6-methoxy-pyridine-3-carbaldehyde, 6-methoxypyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 6-methoxy, 6-methoxy-3-nicotinaldehyde, pubchem5152
PubChem CID 3364576
IUPAC Name 6-methoxypyridine-3-carbaldehyde
SMILES COC1=CC=C(C=O)C=N1

Specifications

Melting Point 49°C to 52°C
Flash Point 107°C (225°F)
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 137.14
Percent Purity 98%
Chemical Name or Material 6-Methoxypyridine-3-carboxaldehyde
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only